Skip to Content
MilliporeSigma
All Photos(1)

Documents

115266

Sigma-Aldrich

4-Bromoisophthalic acid

95%

Synonym(s):

1,3-Dicarboxy-4-bromobenzene, 4-Bromo-1,3-benzenedicarboxylic acid, 4-Bromoisophthalic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H3-1,3-(CO2H)2
CAS Number:
Molecular Weight:
245.03
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

Assay

95%

form

solid

mp

297-299 °C (lit.)

SMILES string

OC(=O)c1ccc(Br)c(c1)C(O)=O

InChI

1S/C8H5BrO4/c9-6-2-1-4(7(10)11)3-5(6)8(12)13/h1-3H,(H,10,11)(H,12,13)

InChI key

MSQIEZXCNYUWHN-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

T L Youngs et al.
Biochemistry international, 23(5), 915-922 (1991-03-01)
To examine the idea that glutamate decarboxylase from E. coli can be a convenient source for the study of the effects of compounds on GABA synthesis in the nervous system, a series of substrate analogues and divalent cations were tested

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service