Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

111147

Sigma-Aldrich

3-Methyl-1-pentene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH2CH(CH3)CH=CH2
CAS Number:
Molecular Weight:
84.16
Beilstein:
1719110
EC Number:
MDL number:
UNSPSC Code:
12352100
eCl@ss:
39010211
PubChem Substance ID:
NACRES:
NA.22

vapor pressure

436 mmHg ( 37.7 °C)

Assay

99%

refractive index

n20/D 1.384 (lit.)

bp

54 °C (lit.)

mp

−154 °C (lit.)

density

0.67 g/mL at 25 °C (lit.)

SMILES string

CCC(C)C=C

InChI

1S/C6H12/c1-4-6(3)5-2/h4,6H,1,5H2,2-3H3

InChI key

LDTAOIUHUHHCMU-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

3-Methyl-1-pentene has been synthesized by the pyrolysis of ethylene –butene-2 mixtures in a static system.

Application

3-Methyl-1-pentene has been used to study the rate constant of chlorine atom.

Pictograms

FlameHealth hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Asp. Tox. 1 - Flam. Liq. 2

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

-16.6 °F - closed cup

Flash Point(C)

-27 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of reactions of chlorine atoms with a series of alkenes at 1 atm and 298 K: structure and reactivity.
Ezell MJ, et al.
Physical Chemistry Chemical Physics, 4(23), 5813-5820 (2002)
Ene reactions of olefins. I. The addition of ethylene to 2-butene and the decomposition of 3-methylpentene-1.
International Journal of Chemical Kinetics, 10(3), 307-324 (1978)
Peter Tieu et al.
Frontiers in chemistry, 8, 599-599 (2020-08-06)
Metal nanoparticle catalysts functionalized with small, well-defined organic ligands are important because such systems can provide a spatial control in the catalyst-substrate interactions. This article describes the synthesis, stability, and catalytic property evaluations of four different Pd nanoparticles capped with

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service