Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

11070

Sigma-Aldrich

Aromadendrene oxide 2

≥95.0%

Sign Into View Organizational & Contract Pricing

Select a Size

100 MG
$200.00

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Select a Size

Change View
100 MG
$200.00

About This Item

Empirical Formula (Hill Notation):
C15H24O
CAS Number:
Molecular Weight:
220.35
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

$200.00


Please contact Customer Service for Availability

Request a Bulk Order

Assay

≥95.0%

functional group

ether

SMILES string

C[C@@H]1CC[C@@H]2[C@@H]1[C@H]3[C@@H](CC[C@@]24CO4)C3(C)C

InChI

1S/C15H24O/c1-9-4-5-10-12(9)13-11(14(13,2)3)6-7-15(10)8-16-15/h9-13H,4-8H2,1-3H3/t9-,10-,11-,12-,13-,15-/m1/s1

InChI key

XPGWKKLDFXNBPJ-QTPLKFIXSA-N

Biochem/physiol Actions

Aromadendrene oxide 2 is one of the biologically active component of Lantana camara that shows antimicrobial property.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service