Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

110507

Sigma-Aldrich

1,4-Dimethylcyclohexane, mixture of cis and trans

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H10(CH3)2
CAS Number:
Molecular Weight:
112.21
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.426 (lit.)

bp

120 °C (lit.)

mp

−87 °C (lit.)

density

0.773 g/mL at 25 °C (lit.)

SMILES string

CC1CCC(C)CC1

InChI

1S/C8H16/c1-7-3-5-8(2)6-4-7/h7-8H,3-6H2,1-2H3

InChI key

QRMPKOFEUHIBNM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1,4-Dimethylcyclohexane (mixture of cis and trans) has been used to study the absorption of ultrasonic waves that can be measured by a reverberation technique.

Signal Word

Danger

Hazard Statements

Hazard Classifications

Aquatic Chronic 2 - Asp. Tox. 1 - Flam. Liq. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Central nervous system

Storage Class Code

3 - Flammable liquids

WGK

WGK 2

Flash Point(F)

60.8 °F - closed cup

Flash Point(C)

16 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service