Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

110272

Sigma-Aldrich

2-Decalone,mixture of cis and trans

95%

Synonym(s):

Decahydro-2-naphthalenone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H16O
CAS Number:
Molecular Weight:
152.23
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

refractive index

n20/D 1.491 (lit.)

bp

96 °C/2.5 mmHg (lit.)

density

0.979 g/mL at 25 °C (lit.)

functional group

ketone

SMILES string

O=C1CCC2CCCCC2C1

InChI

1S/C10H16O/c11-10-6-5-8-3-1-2-4-9(8)7-10/h8-9H,1-7H2

InChI key

LGVJRKCQQHOWAU-UHFFFAOYSA-N

General description

2-Decalone (mixture of cis and trans) is obtained by dosing the extracts of kidney homogenates from male rats with cis- and trans-decalin[1].

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

215.6 °F - closed cup

Flash Point(C)

102 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

C T Olson et al.
Journal of toxicology and environmental health, 18(2), 285-292 (1986-01-01)
The cis- and trans-decalin stereoisomers have found many uses as solvents and fuel components. The metabolism of the decalin isomers in male and female Fischer-344 rats and the effects of the decalins on renal damage were evaluated. Only male rats

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service