Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10927

Sigma-Aldrich

Boc-N-Me-Aib-OH

≥98.0% (HPLC)

Synonym(s):

N-Boc-α-(methylamino)isobutyric acid, Boc-N,2-dimethylalanine, Boc-N-methyl-Aib

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H19NO4
CAS Number:
Molecular Weight:
217.26
Beilstein:
7371082
MDL number:
UNSPSC Code:
12352200
eCl@ss:
32160406
PubChem Substance ID:

Assay

≥98.0% (HPLC)

form

solid

reaction suitability

reaction type: Boc solid-phase peptide synthesis

mp

154-158 °C

application(s)

peptide synthesis

SMILES string

CN(C(=O)OC(C)(C)C)C(C)(C)C(O)=O

InChI

1S/C10H19NO4/c1-9(2,3)15-8(14)11(6)10(4,5)7(12)13/h1-6H3,(H,12,13)

InChI key

KPDLITGXUYMJEC-UHFFFAOYSA-N

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Ryan W Barrs et al.
Chemical reviews, 120(19), 10887-10949 (2020-09-02)
Microvasculature functions at the tissue and cell level, regulating local mass exchange of oxygen and nutrient-rich blood. While there has been considerable success in the biofabrication of large- and small-vessel replacements, functional microvasculature has been particularly challenging to engineer due

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service