Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10890

Sigma-Aldrich

2,5-Hexanedione

produced by Wacker Chemie AG, Burghausen, Germany, ≥97.0% (GC)

Synonym(s):

ACAN, Acetonylacetone

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH2CH2COCH3
CAS Number:
Molecular Weight:
114.14
Beilstein:
506525
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

produced by Wacker Chemie AG, Burghausen, Germany

vapor pressure

0.43 mmHg ( 20 °C)

Assay

≥97.0% (GC)

refractive index

n20/D 1.425 (lit.)

bp

191 °C (lit.)

mp

−6-−5 °C (lit.)

density

0.973 g/mL at 25 °C (lit.)

SMILES string

CC(=O)CCC(C)=O

InChI

1S/C6H10O2/c1-5(7)3-4-6(2)8/h3-4H2,1-2H3

InChI key

OJVAMHKKJGICOG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Other Notes

prices for bulk quantities on request

related product

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service