Skip to Content
MilliporeSigma
All Photos(2)

Documents

107859

Sigma-Aldrich

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride

97%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H7ClN2O3S2
CAS Number:
Molecular Weight:
254.71
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

161-164 °C (lit.)

SMILES string

CC(=O)Nc1nc(C)c(s1)S(Cl)(=O)=O

InChI

1S/C6H7ClN2O3S2/c1-3-5(14(7,11)12)13-6(8-3)9-4(2)10/h1-2H3,(H,8,9,10)

InChI key

NXGKPRKPUCSEIL-UHFFFAOYSA-N

Application

2-Acetamido-4-methyl-5-thiazolesulfonyl chloride was used in high-performance liquid chromatographic (HPLC) assay for acetazolamide.

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

J Gal et al.
Current eye research, 1(6), 361-365 (1981-01-01)
A high-performance liquid chromatographic (HPLC) assay for acetazolamide is presented. A 100-microliter sample is mixed with an aliquot of the internal standard solution and the mixture, buffered at pH 4.5, is extracted with ethyl acetate. The extract is evaporated to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service