Skip to Content
MilliporeSigma
All Photos(1)

Documents

107417

Sigma-Aldrich

2-Amino-1,1,3-propenetricarbonitrile

97%

Synonym(s):

2-Amino-1,1-3-tricyanopropene, Malononitrile dimer

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(NC)2C=C(NH2)CH2CN
CAS Number:
Molecular Weight:
132.12
Beilstein:
1756131
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

171-173 °C (lit.)

SMILES string

N\C(CC#N)=C(\C#N)C#N

InChI

1S/C6H4N4/c7-2-1-6(10)5(3-8)4-9/h1,10H2

InChI key

BNHGNFYPZNDLAF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Biochem/physiol Actions

2-Amino-1,1,3-propenetricarbonitrile is a key intermediate in synthesis of various heterocyclic compounds. 2-Amino-1,1-3-tricyanopropene undergoes condensation reaction with aromatic nitroso compounds to form useful coloring agents. 2-Amino-1,1-3-tricyanopropene reacts with 1,3-dithiole-derived polyenals to form push–pull systems with (Z)-geometry around the newly formed carbon-carbon double bond.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

An improved and facile synthesis of 2-amino-1, 1, 3-tricyanopropene.
Mittelbach M.
Monatshefte fur Chemie / Chemical Monthly, 116(5), 689-691 (1985)
Synthesis, characterization and optical properties of merocyanines derived from malononitrile dimer.
Alias S, et al.
Tetrahedron Letters, 48(37), 6539-6542 (2007)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service