Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

106895

Sigma-Aldrich

1,1′-Ferrocenedicarboxylic acid

96%

Synonym(s):

1,1′-Biscarboxylferrocene, 1,1′-Dicarboxyferrocene

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H10FeO4
CAS Number:
Molecular Weight:
274.05
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

96%

reaction suitability

core: iron
reagent type: catalyst

impurities

<3% ferrocenecarboxylic acid

mp

≥300 °C (lit.)

SMILES string

[Fe].OC(=O)[C]1[CH][CH][CH][CH]1.OC(=O)[C]2[CH][CH][CH][CH]2

InChI

1S/2C6H5O2.Fe/c2*7-6(8)5-3-1-2-4-5;/h2*1-4H,(H,7,8);

InChI key

ISVVAJYTLASNEJ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Reactant for preparation of:
  • Planar chiral ferrocene-based amido-phosphine ligands as catalysts for asymmetric allylic alkylation reactions
  • Potential antitumor agents
  • Isostructural ferrocenyl coordination polymer hollow microspheres with H2 storage properties
  • Prganometallic aromatic polyesters
  • Manganese-lanthanide-ferrocene clusters

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Dongmiao Qin et al.
Mikrochimica acta, 187(8), 482-482 (2020-08-05)
A composite, reduced graphene oxide (rGO) doped with silver nanoparticles (Ag NPs), was prepared by using binary reductants of sodium citrate and hydrazine hydrate. Carbon quantum dots (CQDs) synthesized by papaya peel combined with silver ions to form a CQDs-loaded
Shun Yang et al.
Journal of colloid and interface science, 554, 565-571 (2019-07-22)
Traditional method to functionalize mesoporous silica nanoparticles with organic groups for removal of contaminants from wastewater was surface modification. However, this surface modification could not cover the entire surface, leading to incomplete utilization of the high surface area of MSNs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service