Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

10635

Sigma-Aldrich

9,10-Anthraquinone-2,6-disulfonic acid disodium salt

purum, ≥97.0% (HPLC)

Synonym(s):

9,10-Dihydro-9,10-dioxo-2,6-anthracenedisulfonic acid disodium salt, Anthraquinone-2,6-disulfonic acid disodium salt, Disodium anthraquinone-2,6-disulfonate

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H6Na2O8S2
CAS Number:
Molecular Weight:
412.30
Beilstein:
3582949
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

grade

purum

Assay

≥97.0% (HPLC)

impurities

≤3% water

solubility

H2O: 10 mg/mL, clear

SMILES string

[Na+].[Na+].[O-]S(=O)(=O)c1ccc2C(=O)c3cc(ccc3C(=O)c2c1)S([O-])(=O)=O

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service