Skip to Content
MilliporeSigma
All Photos(1)

Documents

103985

Sigma-Aldrich

Cyclopentanemethanol

98%

Synonym(s):

(Hydroxymethyl)cyclopentane, Cyclopentyl carbinol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C5H9CH2OH
CAS Number:
Molecular Weight:
100.16
Beilstein:
1919000
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.458 (lit.)

bp

162-163 °C (lit.)

density

0.926 g/mL at 25 °C (lit.)

SMILES string

OCC1CCCC1

InChI

1S/C6H12O/c7-5-6-3-1-2-4-6/h6-7H,1-5H2

InChI key

ISQVBYGGNVVVHB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Cyclopentanemethanol was used to study catalytic reduction of 6-bromo-1-hexene by nickel (I) salen electrogenerated at a glassy carbon electrode in acetonitrile containing tetramethylammonium tetrafluoroborate by cyclic voltammetry and controlled-potential electrolysis.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

143.6 °F - closed cup

Flash Point(C)

62 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Catalytic reduction of 6-bromo-1-hexene by nickel (I) salen electrogenerated at glassy carbon cathodes in acetonitrile.
Fang DM, et al.
Journal of the Electrochemical Society, 148(12), E464-E467 (2001)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service