Skip to Content
MilliporeSigma
All Photos(1)

Key Documents

102709

Sigma-Aldrich

N-Ethyl-1-naphthylamine

98%

Synonym(s):

α-Ethylaminonaphthalene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C10H7NHC2H5
CAS Number:
Molecular Weight:
171.24
Beilstein:
2207404
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

refractive index

n20/D 1.644 (lit.)

bp

175-176 °C/15 mmHg (lit.)

density

1.065 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

CCNc1cccc2ccccc12

InChI

1S/C12H13N/c1-2-13-12-9-5-7-10-6-3-4-8-11(10)12/h3-9,13H,2H2,1H3

InChI key

KDFFXYVOTKKBDI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

N-Ethyl-1-naphthylamine was used in characterization of nitrite-dependent genotoxins and tumour promoter-like substances in fermented fish sauce by cross coupling reaction. It may be used in the identification of triazenes metabolites in urine by coupling and forming colored azo dyes.

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Aquatic Acute 1

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metabolism of the Tumour-Inhibitory 3.3-Dimethyl-1-Phenyl-Triazene and Its 4-Chlorophenyl Analogue.
Kolar GF and Schlesiger J.
Chemotherapy, 91-96 (1976)
C S Chen et al.
Mutation research, 265(2), 211-221 (1992-02-01)
A high gastric cancer mortality in Fujian province (Peoples Republic of China) has been associated with the consumption of certain salted fermented fish products such as fish sauce (FS). We have investigated the levels and nature of N-nitroso compounds (NOC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service