Skip to Content
MilliporeSigma
All Photos(1)

Documents

102598

Sigma-Aldrich

3,3′-Dimethoxybenzidine

97%

Synonym(s):

o-Dianisidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[-C6H3(OCH3)NH2]2
CAS Number:
Molecular Weight:
244.29
Beilstein:
1879884
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

97%

mp

137-138 °C (lit.)

SMILES string

COc1cc(ccc1N)-c2ccc(N)c(OC)c2

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

S Torres-Cartas et al.
European journal of medicinal chemistry, 42(11-12), 1396-1402 (2007-05-08)
Mutagenicity is a toxicity endpoint associated with the chronic exposure to chemicals. Aromatic amines have considerable industrial and environmental importance due to their widespread use in industry and their mutagenic capacity. Biopartitioning micellar chromatography (BMC), a mode of micellar liquid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service