Skip to Content
MilliporeSigma
All Photos(1)

Documents

07430

Sigma-Aldrich

2-Chloro-5-(trifluoromethyl)aniline

≥98.0% (GC)

Synonym(s):

3-Amino-4-chlorobenzotrifluoride, 6-Chloro-α,α,α-trifluoro-m-toluidine

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
ClC6H3(CF3)NH2
CAS Number:
Molecular Weight:
195.57
Beilstein:
879910
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

≥98.0% (GC)

refractive index

n20/D 1.499 (lit.)

density

1.428 g/mL at 25 °C (lit.)

SMILES string

Nc1cc(ccc1Cl)C(F)(F)F

InChI

1S/C7H5ClF3N/c8-5-2-1-4(3-6(5)12)7(9,10)11/h1-3H,12H2

InChI key

VKTTYIXIDXWHKW-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation markEnvironment

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

167.0 °F - closed cup

Flash Point(C)

75 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service