- Structure/activity relationship of thapsigargin inhibition on the purified Golgi/secretory pathway Ca
Structure/activity relationship of thapsigargin inhibition on the purified Golgi/secretory pathway Ca
The Journal of biological chemistry (2017-03-08)
Jialin Chen, Joren De Raeymaecker, Jannik Brøndsted Hovgaard, Susanne Smaardijk, Ilse Vandecaetsbeek, Frank Wuytack, Jesper Vuust Møller, Jan Eggermont, Marc De Maeyer, Søren Brøgger Christensen, Peter Vangheluwe
PMID28264934
ABSTRACT
The Golgi/secretory pathway Ca