Skip to Content
Merck
CAPSO ≥99% anhydrous basis (titration)

CAPSO

CAPSO ≥99% anhydrous basis (titration)
Synonym(s):
3-(Cyclohexylamino)-2-hydroxy-1-propanesulfonic acid, CAPSO Free Acid
Empirical Formula (Hill Notation):
C9H19NO4S
CAS Number:
Molecular Weight:
237.32

Products

Product Category
Brand
Feature
Formula Weight
pH Value
Physical Form
pK Value
Purity
Quality Segment
Application
Special Grade
Showing 1-1 of 1
Compare
Product Number
Description
Pricing