Skip to Content
Merck

Skip To

1605512

USP

Ropivacaine Related Compound A

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2,6-Dimethylaniline hydrochloride, 2,6-Xylidine hydrochloride, 2,6-Xylidinium chloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C8H11N · HCl
CAS Number:
Molecular Weight:
157.64
MDL number:
UNSPSC Code:
41116107
NACRES:
NA.24
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

ropivacaine

manufacturer/tradename

USP

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

[Cl-].[N+H3]c1c(cccc1C)C

InChI

1S/C8H11N.ClH/c1-6-4-3-5-7(2)8(6)9;/h3-5H,9H2,1-2H3;1H

InChI key

QNWMFJDRMZWZNN-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Ropivacaine Related Compound A USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
PHR2572PHR23061605523
grade

pharmaceutical primary standard

grade

certified reference material, pharmaceutical secondary standard

grade

-

grade

pharmaceutical primary standard

manufacturer/tradename

USP

manufacturer/tradename

-

manufacturer/tradename

-

manufacturer/tradename

USP

format

neat

format

-

format

-

format

neat

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical small molecule

application(s)

pharmaceutical (small molecule)

API family

ropivacaine

API family

ropivacaine

API family

ropivacaine

API family

ropivacaine


Still not finding the right product?

Explore all of our products under Ropivacaine Related Compound A


Storage Class

11 - Combustible Solids

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters