Skip to Content
Merck

1457403

USP

Naproxen sodium

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

(S)-6-Methoxy-α-methyl-2-naphthaleneacetic acid sodium salt

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C14H13NaO3
CAS Number:
Molecular Weight:
252.24
NACRES:
NA.24
PubChem Substance ID:
UNSPSC Code:
41116107
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

naproxen

manufacturer/tradename

USP

application(s)

pharmaceutical
pharmaceutical (small molecule)

format

neat

SMILES string

[Na+].COc1ccc2cc(ccc2c1)[C@H](C)C([O-])=O

InChI

1S/C14H14O3.Na/c1-9(14(15)16)10-3-4-12-8-13(17-2)6-5-11(12)7-10;/h3-9H,1-2H3,(H,15,16);/q;+1/p-1/t9-;/m0./s1

InChI key

CDBRNDSHEYLDJV-FVGYRXGTSA-M

Gene Information

General description

Find answers to frequently asked questions on USP products in USP FAQs.
This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Naproxen sodium USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Naproxen Sodium and Pseudoephedrine Hydrochloride Extended-Release Tablets
  • Naproxen Sodium Tablets

Biochem/physiol Actions

Cyclooxygenase (Prostaglandin H synthase 1 and 2) inhibitor.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.


Still not finding the right product?

Explore all of our products under Naproxen sodium


Pictograms

Health hazardExclamation mark

Signal Word

Danger

Hazard Codes

Hazard Classifications

Acute Tox. 4 Oral - Repr. 1A

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library