Skip to Content
Merck

Skip To

1300004

USP

Guaiacol

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

2-Methoxyphenol, Catechol monomethyl ether, Pyrocatechol monomethyl ether

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Linear Formula:
(CH3O)C6H4OH
CAS Number:
Molecular Weight:
124.14
UNSPSC Code:
41116107
NACRES:
NA.24
PubChem Substance ID:
MDL number:
Beilstein/REAXYS Number:
508112
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

vapor density

4.27 (vs air)

vapor pressure

0.11 mmHg ( 25 °C)

API family

guaiacol

manufacturer/tradename

USP

refractive index

n20/D 1.543 (lit.)

bp

205 °C (lit.)

mp

26-29 °C (lit.)

density

1.129 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

storage temp.

2-8°C

SMILES string

COc1ccccc1O

InChI

1S/C7H8O2/c1-9-7-5-3-2-4-6(7)8/h2-5,8H,1H3

InChI key

LHGVFZTZFXWLCP-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Biochem/physiol Actions

Guaiacol, along with 2,4,6-trichloroanisole, is responsible for cork taint in wine. A method has been developed for extraction and quantitation of the two compounds.

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
Y0000619G550212-1620
grade

pharmaceutical primary standard

grade

pharmaceutical primary standard

grade

-

grade

SAJ first grade

manufacturer/tradename

USP

manufacturer/tradename

EDQM

manufacturer/tradename

-

manufacturer/tradename

-

format

neat

format

neat

format

-

format

-

application(s)

pharmaceutical (small molecule)

application(s)

pharmaceutical (small molecule)

application(s)

-

application(s)

-

mp

26-29 °C (lit.)

mp

26-29 °C (lit.)

mp

26-29 °C (lit.)

mp

26-29 °C (lit.)

vapor pressure

0.11 mmHg ( 25 °C)

vapor pressure

0.11 mmHg ( 25 °C)

vapor pressure

0.11 mmHg ( 25 °C)

vapor pressure

0.11 mmHg ( 25 °C)


Still not finding the right product?

Explore all of our products under Guaiacol


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters