Skip to Content
Merck

1200804

USP

Dihydrocodeine bitartrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Dihydrocodeine hydrogen tartrate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C18H23NO3 · C4H6O6
CAS Number:
Molecular Weight:
451.47
MDL number:
UNSPSC Code:
41116107
EC Number:
227-747-7
NACRES:
NA.24
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


grade

pharmaceutical primary standard

agency

USP

API family

codeine

manufacturer/tradename

USP

drug control

USDEA Schedule II; regulated under CDSA - not available from Sigma-Aldrich Canada; estupefaciente (Spain); Decreto Lei 15/93: Tabela IA (Portugal), kontrollierte Droge in Deutschland

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

N1(C2C3C4(C(Oc5c4c(ccc5OC)C2)C(CC3)O)CC1)C.OC(C(O)C(=O)O)C(=O)O

InChI

1S/C18H23NO3.C4H6O6/c1-19-8-7-18-11-4-5-13(20)17(18)22-16-14(21-2)6-3-10(15(16)18)9-12(11)19;5-1(3(7)8)2(6)4(9)10/h3,6,11-13,17,20H,4-5,7-9H2,1-2H3;1-2,5-6H,(H,7,8)(H,9,10)

InChI key

ZGSZBVAEVPSPFM-UHFFFAOYSA-N

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.
For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Dihydrocodeine bitartrate USP Reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Hydrocodone Bitartrate and Homatropine Methylbromide Tablets
  • Hydrocodone Bitartrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.
USDEA Schedule 2 Item

Legal Information

German
Dieses Produkt fällt unter das Betäubungsmittelgesetz (BtMG). Für eine Bestellung dieses Produktes ist eine Erlaubnis nach § 3 BtMG zwingend erforderlich, es sei denn, es greift eine Ausnahme von der Erlaubnispflicht nach § 4 oder § 26 BtMG.

English
This product is subject to the German Narcotics Act. A permit under Section 3 of the German Narcotics Act is mandatory for ordering this product unless an exemption from the permit requirement under Section 4 or Section 26 of the German Narcotics Act applies.


Still not finding the right product?

Explore all of our products under Dihydrocodeine bitartrate


pictograms

Exclamation mark

signalword

Warning

hcodes

Hazard Classifications

Acute Tox. 4 Oral - STOT SE 3

target_organs

Central nervous system

Storage Class

11 - Combustible Solids

wgk

WGK 1

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library