Skip to Content
Merck
All Photos(1)

Key Documents

808029

TSKgel® G2500PW (12 µm) HPLC Columns

L x I.D. 60 cm × 7.5 mm, hydroxylated methacrylate phase

Synonym(s):

TSKgel® Size Exclusion (PW-Type) HPLC Column

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41115711
eCl@ss:
32110501

product name

TSKgel® G2500PW HPLC Column, phase hydroxylated methacrylate, L × I.D. 60 cm × 7.5 mm, 12 μm particle size

material

stainless steel hardware

Agency

suitable for USP L25
suitable for USP L38
suitable for USP L39

product line

TSKgel®

manufacturer/tradename

Tosoh 08029

parameter

≤0.5 M salt concentration
≤20% organic solvent
1.2 mL/min flow rate
40 bar max. pressure
80 °C temperature

technique(s)

HPLC: suitable

L × I.D.

60 cm × 7.5 mm

matrix

polymer particle platform

matrix active group

hydroxylated methacrylate phase

particle size

12 μm

pore size

<200 Å

operating pH

2-12

separation technique

size exclusion (SEC)

Looking for similar products? Visit Product Comparison Guide

General description

Use TSKgel® G2500PW for the analysis of water-soluble polymers with molecular weights of less than 3,000 Da.

Physical form

Shipped in water.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

TSKgel is a registered trademark of Tosoh Corporation

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service