Skip to Content
Merck
All Photos(1)

Key Documents

64822-U

Supelco

Flat Bottom Flask

size 250 mL

Synonym(s):

FLAT BOTTOM FLASK,250ML,24/40, EA

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12000000
NACRES:
NB.21

material

flat bottom glass flask

size

medium
250 mL

Joint

joint ST/NS flask

application(s)

food and beverages

Looking for similar products? Visit Product Comparison Guide

Related Categories


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service