Skip to Content
Merck
All Photos(2)

Key Documents

57100-U

Supelco

Visidry Drying Attachment

for use with Visiprep 12-port model

Synonym(s):

SPE Vacuum Manifold Drying Accessory

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121800
NACRES:
NB.21

Agency

suitable for DIN 38407-42
suitable for EPA 1633
suitable for EPA 533
suitable for EPA 537.1
suitable for EPA OTM-45
suitable for GB 31604.35-2016
suitable for ISO 21675 2019
suitable for ISO 25101
suitable for ISO/CEN 15968-2010

reg. compliance

suitable for FDA C-010.02

technique(s)

solid phase extraction (SPE): suitable

application(s)

food and beverages

compatibility

for use with Visiprep 12-port model

Looking for similar products? Visit Product Comparison Guide

General description

The Visidry Drying Attachment was designed for our Visiprep Vacuum Manifold (Cat. No. 57100-U also fits our economical Preppy manifold). The Visidry unit installs in minutes, dries up 12 or up to 24 SPE tubes at one time, and can be used with any inert gas supply. It is also useful for evaporating and concentrating recovered samples. Gas flow to each port can be independently adjusted.

Note: The Visidry drying attachment cannot be used to dry 12mL, 20mL, or 60mL SPE tubes.

Legal Information


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Related Content

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

FAS, or 'forever chemicals,' persist in the environment and pose risks to human health. Discover our environmental monitoring tools for PFAS quantification, aiding researchers, regulators, and labs in testing for PFAS.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service