Skip to Content
Merck
All Photos(1)

Key Documents

44718

Supelco

3-Ethoxy-1,2-propanediol

for GC derivatization, ≥99.0% (GC)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C2H5OCH2CH(OH)CH2OH
CAS Number:
Molecular Weight:
120.15
MDL number:
UNSPSC Code:
12191601
NACRES:
NA.21

grade

derivatization grade ((GC derivatization))
for GC derivatization

Quality Level

Assay

≥99.0% (GC)

refractive index

n20/D 1.441 (lit.)

bp

222 °C (lit.)

density

1.063 g/mL at 25 °C (lit.)

SMILES string

CCOCC(O)CO

InChI

1S/C5H12O3/c1-2-8-4-5(7)3-6/h5-7H,2-4H2,1H3

InChI key

LOSWWGJGSSQDKH-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application


  • Matrix Enhancement Effect in Gas Chromatography: Explores the utility of 3-Ethoxy-1,2-propanediol as an analyte protectant to mitigate matrix effects in gas chromatographic analysis, enhancing the reliability of pesticide residue detection and other complex sample analyses in environmental science and analytical chemistry (Rahman et al., 2013).

  • Automated Direct Sample Introduction in GC-MS: Discusses the integration of 3-Ethoxy-1,2-propanediol in automated systems for pesticide residue analysis using GC-MS, highlighting its role in improving sample introduction efficiency and accuracy, crucial for lab technicians and researchers in environmental and food safety fields (Cajka et al., 2005).

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Skin Irrit. 2

Storage Class Code

10 - Combustible liquids

WGK

WGK 2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service