ZRB1403
Anti-p-4E-BP1 (Thr37/46) Antibody, clone 1N7 ZooMAb® Rabbit Monoclonal

recombinant, expressed in HEK 293 cells
About This Item
Recommended Products
biological source
rabbit
Quality Level
recombinant
expressed in HEK 293 cells
conjugate
unconjugated
antibody form
purified antibody
antibody product type
primary antibodies
clone
1N7, recombinant monoclonal
description
recombinant, expressed in HEK 293 cells
product line
ZooMAb® learn more
form
lyophilized
mol wt
calculated mol wt 12.58 kDa
observed mol wt ~15 kDa
purified by
using Protein A
species reactivity
mouse, human
species reactivity (predicted by homology)
feline, porcine, monkey, canine, bovine
packaging
antibody small pack of 25 μL
greener alternative product characteristics
Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.
enhanced validation
recombinant expression
Learn more about Antibody Enhanced Validation
sustainability
Greener Alternative Product
technique(s)
affinity binding assay: suitable
immunocytochemistry: suitable
immunohistochemistry (formalin-fixed, paraffin-embedded sections): suitable
western blot: suitable
isotype
IgG
epitope sequence
Unknown
Protein ID accession no.
UniProt accession no.
greener alternative category
shipped in
ambient
storage temp.
2-8°C
target post-translational modification
phosphorylation (pThr37/pThr46)
General description
Specificity
Immunogen
Application
Evaluated by Western Blotting in BALB/3T3 A31 cell lysate.
Western Blotting Analysis (WB): A 1:10,000 dilution of this antibody detected 4E-BP1 phosphorylated on Threonine 37 and 46 in lysate from overnight serum starved and PDGF treated ( 50 ng/mL) Balb/3T3 A31 cells.
Tested applications
Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected p-4E-BP1 (Thr37/46) in lysate from NIH3T3 cells stimulated with PDGF (50 ng/mL)
Immunohistochemistry (Paraffin) Analysis: A 1:1,000 dilution from a representative lot detected p-4E-BP1 (Thr37/46) in human pancreas and human colon cancer tissue sections.
Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected p-4E-BP1 (Thr37/46) in NIH 3T3 cells with and without PDGF treatment.
Peptide Inhibition Assay: Target band detection in lysate from NIH3T3 cells treated with PDGF was prevented by preblocking of a representative lot with the immunogen phosphopeptide ( p-4E-BP1), but not the corresponding non-phosphopeptide.
Affinity Binding Assay: A representative lot of this antibody bound phospho and non-phospho 4E-BP1-Thr37+46 peptides with a KD of 1x 10^-12 in an affinity binding assay.
Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Target description
Physical form
Storage and Stability
Legal Information
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service