Skip to Content
Merck
All Photos(9)

Key Documents

ZRB1183

Sigma-Aldrich

Anti-RFX7 Antibody, clone 1J22 ZooMAb® Rabbit Monoclonal

enhanced validation
greener alternative

recombinant, expressed in HEK 293 cells

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

Quality Level

recombinant

expressed in HEK 293 cells

conjugate

unconjugated

antibody form

purified antibody

antibody product type

primary antibodies

clone

1J22, monoclonal
recombinant monoclonal

product line

ZooMAb® learn more

form

lyophilized

mol wt

calculated mol wt ~146.90 kDa
observed mol wt ~200 kDa

species reactivity

human

packaging

antibody small pack of 25 μL

greener alternative product characteristics

Waste Prevention
Designing Safer Chemicals
Design for Energy Efficiency
Learn more about the Principles of Green Chemistry.

enhanced validation

recombinant expression
Learn more about Antibody Enhanced Validation

sustainability

Greener Alternative Product

technique(s)

affinity binding assay: suitable
immunocytochemistry: suitable
western blot: suitable

isotype

IgG

UniProt accession no.

greener alternative category

shipped in

ambient

storage temp.

2-8°C

target post-translational modification

unmodified

General description

We are committed to bringing you greener alternative products, which adhere to one or more of The 12 Principles of Green Chemistry.This antibody is Preservative-free, produced without the harm or sacrifice of animals and exceptionally stable to allow for ambient shipping and storage if needed and thus aligns with "Waste Prevention", "Designing Safer Chemicals" and "Design for Energy Efficiency". Click here for more information.
ZooMAb antibodies represent an entirely new generation of recombinant monoclonal antibodies.

Each ZooMAb antibody is manufactured using our proprietary recombinant expression system, purified to homogeneity, and precisely dispensed to produce robust and highly reproducible lot-to-lot consistency. Only top-performing clones are released for use by researchers. Each antibody is validated for high specificity and affinity across multiple applications, including its most commonly used application. ZooMAb antibodies are reliably available and ready to ship when you need them.

Specificity

Clone 1J22 is a ZooMAb® rabbit recombinant monoclonal antibody that specifically detects DNA-binding protein RFX7. It targets an epitope within the N-terminal half.

Immunogen

His-tagged recombinant fragment corresponding to 250 amino acids from the N-terminal half of human DNA-binding protein RFX7.

Application

Quality Control Testing

Evaluated by Western Blotting in U2OS cell nuclear extract.

Western Blotting Analysis: A 1:1,000 dilution of this antibody detected RFX7 in U2OS cell nuclear extract.

Tested Applications

Western Blotting Analysis: A 1:1,000 dilution from a representative lot detected RFX7 in HeLa cell nuclear extract.

Immunocytochemistry Analysis: A 1:100 dilution from a representative lot detected RFX7 in A431 cells.

Affinity Binding Assay: A representative lot of this antibody bound RFX7 proteins with a KD of 8.4 x 10-7 in an affinity binding assay.

Note: Actual optimal working dilutions must be determined by end user as specimens, and experimental conditions may vary with the end user
Anti-RFX7, clone 1J22 ZooMAb®, Cat. No. ZRB1183, is a rabbit recombinant monoclonal antibody that specifically targets RFX7 and is tested for use in Affinity Binding Assay, Immunocytochemistry, and Western Blotting.

Target description

DNA-binding protein RFX7 (UniProt: Q2KHR2; also known as Regulatory factor X 7, Regulatory factor X domain-containing protein 2) is encoded by the RFX7 (also known as RFXDC2) gene (Gene ID: 64864) in human. RFX7 is a member of RFX family of transcription factors that have seven known members. RFX transcription factors are important for development and are involved in the pathogenesis of ciliopathies. They are highly conserved and share a winged-helix DNA-binding domain (DBD) recognizing X-box motifs. RFX7 is expressed in many different tissues examined, with highest levels observed in the brain. Deletion of RFX7 gene leads to a decrease in natural killer cell maintenance and immunity and knockdown of RFX7 in xenopus embryo is shown to result in defects in ciliogenesis in the neural tube and failure of neural tube closure. RFX7 contains an PXLPXL/I motif (aa 91-96) that mediates interaction with Ankyrin Repeat Family A Member 2 (ANKRA2) and its homolog, Regulatory Factor X Associated Ankyrin Containing Protein (RFXANK). This ZooMAb® recombinant monoclonal antibody, generated by our propriety technology, offers significantly enhanced specificity, affinity, reproducibility, and stability over conventional monoclonals. Castro, W., et al. (2018). Nat. Immunol. 19(8); 809-820; Manojlovic, Z., et al. (2014). Mech, Dev. 132(1); 28-37; Gao, J., and Xu, C (2020). Biochem. Biophys. Res. Commun. 523(1); 263-266).

Physical form

Purified recombinant mouse monoclonal antibody IgG, lyophilized in PBS, 5% Trehalose, normal appearance a coarse or translucent resin. Contains no biocide or preservatives, such as azide, or any animal by-products. Larger pack sizes provided as multiples of 25 μL.

Storage and Stability

Recommend storage of lyophilized product at 2-8°C; Before reconstitution, micro-centrifuge vials briefly to spin down material to bottom of the vial; Reconstitute each vial by adding 25 μL of filtered lab grade water or PBS; Reconstituted antibodies can be stored at 2-8°C, or -20°C for long term storage. Avoid repeated freeze-thaws.

Legal Information

ZooMAb is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

11 - Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service