Z325953
BRAND® BLAUBRAND® ETERNA graduated pipette, calibrated to deliver
type 3, (zero on top), capacity 25 mL , with 0.1 subdivisions
About This Item
Recommended Products
material
AR-Glas®
Agency
according to DIN 12 697
feature
class AS with batch number
graduation
subdivision 0.1 mL
with large suction tube suitable for cotton plugs
packaging
pack of 6
manufacturer/tradename
BRAND 28415
measuring range
accuracy: 0.1 mL
capacity
25 mL , with 0.1 subdivisions
Looking for similar products? Visit Product Comparison Guide
General description
Features and Benefits
- BRAND® BLAUBRAND® ETERNA 25 mL graduated pipettes allow for easy and accurate volumetric measurement of liquids.
- Manufactured from high-quality materials that meet the stringent laboratory requirements.
- Optimized flow geometry in the tip ensures reliable liquid dispensing.
- The mechanical resilience of the tip and its fire-polished edges enables a long service life.
- Calibrated to delivery with 5 seconds waiting time.
- Reduced waiting time and simplified handling.
- Set the meniscus only once at partial volume.
- Amber stain clear inscriptions on the pipettes include conformity certification, nominal capacity, error limit, reference temperature, calibration ′Ex′, batch/serial number, and volumetric markings.
Other Notes
Legal Information
Choose from one of the most recent versions:
Certificates of Analysis (COA)
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Protocols
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
This page contains detailed information on working with and handling pipettes and pipetting aids.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service