T1918
Nalgene® centrifuge tubes, Oak Ridge Style 3118
nominal capacity 50 mL
Synonym(s):
High-Speed Centrifuge Tubes
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41121703
eCl@ss:
32119210
NACRES:
NB.11
Recommended Products
material
polypropylene screw closure
round bottom tube
screw top cap
transparent polycarbonate
sterility
non-sterile
feature
RCF 50,000 × g
packaging
pack of 10
manufacturer/tradename
Nalgene® 3118-0050
nominal capacity
50 mL
Looking for similar products? Visit Product Comparison Guide
Related Categories
Features and Benefits
- Can withstand temperatures ranging from -135°C to +135°C (-211°F to +275°F).
- Designed with a round bottom to fit high-speed rotors.
- uitable for use in both refrigerated and non-refrigerated centrifuges.
- Includes polypropylene linerless closures, which are easy to clean and provide leakproof protection under normal conditions up to 10,000 x g.
- The tubes are autoclavable.
Legal Information
Nalgene is a registered trademark of Thermo Fisher Scientific or its subsidiaries
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service