Skip to Content
Merck

Skip To

T1753

Trizma® Pre-Set Crystals

Type 7.4-FT, pH 7.4, suitable for use with fresh water species

Synonym(s):

Tris crystals

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

NACRES:
NA.25
UNSPSC Code:
12161700
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


type

Type 7.4-FT

Quality Segment

form

crystalline powder

pH

7.4

solubility

water: 0.66 g/mL, clear, colorless to faintly yellow

suitability

suitable for use with fresh water species

Application

For use with fresh water species.

Legal Information

Trizma is a registered trademark of Merck KGaA, Darmstadt, Germany

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
T1878T7818T7943
Quality Level

200

Quality Level

200

Quality Level

200

Quality Level

200

form

crystalline powder

form

crystalline powder

form

crystalline powder

form

crystalline powder

pH

7.4

pH

8.3

pH

7.50 (25 °C, 0.1 M)(lit.)

pH

7.60 (25 °C, 0.1 M)(lit.)

solubility

water: 0.66 g/mL, clear, colorless to faintly yellow

solubility

water: 0.66 g/mL, clear, colorless

solubility

water: 0.66 g/mL, clear, colorless

solubility

water: 0.66 g/mL, clear, colorless

suitability

suitable for use with fresh water species

suitability

suitable for use with marine species

suitability

-

suitability

-

type

Type 7.4-FT

type

Type 8.3-FT

type

-

type

-


Still not finding the right product?

Explore all of our products under Trizma® Pre-Set Crystals


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

target_organs

Respiratory system

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable

ppe

dust mask type N95 (US), Eyeshields, Gloves



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters