SYNF44243L
Synthware™ three neck round bottom flask with vertical side necks
bottle capacity 3000 mL, side joint: ST/NS 24/40
Synonym(s):
Synthware F44243L, 3-neck round flask
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
UNSPSC Code:
41104812
NACRES:
NB.43
Recommended Products
material
clear
Joint
side joint: ST/NS24/40
packaging
pack of 1 ea
manufacturer/tradename
Kemtech F44243L
bottle capacity
3000 mL
General description
Flask, Round Bottom, 3 Neck, Center 24/40, Side 24/40, 3L
Application
kemetch; synthware; flask; round bottom; three neck
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service