SRP5242
ERK5, active, GST tagged human
PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution
Synonym(s):
BMK1, ERK4, MAPK7, PRKM7
Sign Into View Organizational & Contract Pricing
All Photos(2)
About This Item
Recommended Products
biological source
human
recombinant
expressed in baculovirus infected Sf9 cells
product line
PRECISIO® Kinase
Assay
≥70% (SDS-PAGE)
form
buffered aqueous glycerol solution
specific activity
9.4-12.7 nmol/min·mg
mol wt
~132 kDa
NCBI accession no.
application(s)
cell analysis
shipped in
dry ice
storage temp.
−70°C
Gene Information
human ... MAPK7(5598)
Physical form
Supplied in 50mM Tris-HCl, pH 7.5, 150mM NaCl, 10mM glutathione, 0.1mM EDTA, 0.25mM DTT, 0.1mM PMSF, 25% glycerol.
Preparation Note
after opening, aliquot into smaller quantities and store at -70 °C. Avoid repeating handling and multiple freeze/thaw cycles
Legal Information
PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany
Storage Class Code
10 - Combustible liquids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
MAPK7 gene controls proliferation, migration and cell invasion in osteosarcoma.
Tesser-Gamba F
Molecular Carcinogenesis (2016)
C1q-TNF-related protein-9 promotes cardiac hypertrophy and failure
Mahesh Appari
Circulation Research (2016)
ERK5 Mediated Signalling in Diabetic Retinopathy
Yuexiu Wu
Medical Hypotheses (2015)
Role of MAPK7 in cell proliferation and metastasis in ovarian cancer
Jinhua D
International Journal of Clinical and Experimental Pathology (2015)
Erk5 null mice display multiple extraembryonic vascular and embryonic cardiovascular defects
Christopher
Proceedings of the National Academy of Sciences of the USA (2002)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service