Skip to Content
Merck
All Photos(1)

Documents

SRP3250

Sigma-Aldrich

Oncostatin-M from rat

recombinant, expressed in E. coli, ≥98% (SDS-PAGE), ≥98% (HPLC), suitable for cell culture

Synonym(s):

OSM

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352202
NACRES:
NA.32

biological source

rat

recombinant

expressed in E. coli

Assay

≥98% (HPLC)
≥98% (SDS-PAGE)

form

lyophilized

mol wt

24.4 kDa

packaging

pkg of 10 μg

technique(s)

cell culture | mammalian: suitable

impurities

<0.1 EU/μg endotoxin, tested

color

white to off-white

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

Gene Information

rat ... OSM(289747)

General description

OSM (oncostatin M) is a polypeptide growth factor that belongs to the interleukin-6 cytokine family that includes interleukin-6, interleukin-11, leukemia inhibitory factor, ciliary neurotrophic factor, cardiotrophin-1, and novel neutrophin-1/B-cell-stimulating factor-3. In rats, this protein shows high expression in the late fetal and neonatal testis. It is also expressed at lower levels in the maturing and adult testis. It is also thought to be expressed in Leydig cells in developing testis.
Recombinant rat Oncostatin M is a 24.4kDa protein, containing 215 amino acid residues.

Biochem/physiol Actions

OSM (oncostatin M) functions as a ligand for two high affinity heterodimer receptors that share the signal transducing receptor gp130 (glycoprotein). The two receptors are- type I OSM receptor which is a dimer of the gp130 transducing receptor and the LIF (leukocyte inhibitory factor) receptor β-subunit, and the type II OSM receptor which contains a dimer of gp130 and the OSM receptor β-subunit. OSM is produced by neonatal Sertoli cells, where it induces Sertoli cell and gonocyte proliferation. It is responsible for the induction of spermatogenesis, and negates the effect of luteinizing hormone (LH) on Leydig cell progenitor formation, thus, acting as a regulator for this process. Study in rat shows that this protein stimulates the differentiation of OC15-5 (oval cells) into hepatocytes, and therefore, is thought to be key in liver regeneration by promoting oval cell differentiation into hepatocytes.

Sequence

MKRGCSSSSP KLLSQLKSQA NITGNTASLL EPYILHQNLN TLTLRAACTE HPVAFPSEDM LRQLSKPDFL STVHATLGRV WHQLGAFRQQ FPKIQDFPEL ERARQNIQGI RNNVYCMARL LHPPLEIPEP TQADSGTSRP TTTAPGIFQI KIDSCRFLWG YHRFMGSVGR VFEEWGDGSR RSRRHSPLWA WLKGDHRIRP SRSSQSAMLR SLVPR

Physical form

Lyophilized with no additives.

Reconstitution

Centrifuge the vial prior to opening. Reconstitute in water to a concentration of 0.1-1.0 mg/ml. Do not vortex. This solution can be stored at 2-8°C for up to 1 week. For extended storage, it is recommended to further dilute in a buffer containing a carrier protein (example 0.1% BSA) and store in working aliquots at -20°C to -80°C.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Oncostatin-M inhibits luteinizing hormone stimulated Leydig cell progenitor formation in vitro.
Teerds KJ et al
Reproductive Biology and Endocrinology, 5, 43-43 (2007)
Oncostatin M inhibits proliferation of rat oval cells, OC15-5, inducing differentiation into hepatocytes.
Okaya A et al
The American Journal of Pathology, 166(3), 709-719 (2005)
Bruce, A.G., et al.
Cytokine Reference, 585-598 (2001)
Zhen Wang et al.
Molecular medicine reports, 19(3), 1529-1542 (2018-12-29)
Psoriasis is a chronic inflammatory skin disease. Keratinocytes (KCs), as skin‑specific cells, serve an important role in the immunopathogenesis of psoriasis. In the present study, transcriptome data derived from psoriasis‑like KCs were used together with the reported transcriptome data from
T M Rose et al.
Proceedings of the National Academy of Sciences of the United States of America, 88(19), 8641-8645 (1991-10-01)
Oncostatin M (OSM), a glycoprotein of Mr approximately 28,000 produced by activated monocyte and T-lymphocyte cell lines, was previously identified by its ability to inhibit the growth of cells from melanoma and other solid tumors. We have detected significant similarities

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service