Skip to Content
Merck
All Photos(1)

Key Documents

SML3861

Sigma-Aldrich

Methoxy-X04

≥98% (HPLC)

Synonym(s):

4,4′-(1E,1′E)-2,2′-(2-Methoxy-1,4-phenylene)bis(ethene-2,1-diyl)diphenol, 4,4′-[(2-Methoxy-1,4-phenylene)di-(1E)-2,1-ethenediyl]bisphenol, MX04, MXO4, MeO-X04, MeO-XO4, MeX04, MeXO4, Methoxy-XO4, MethoxyX04, MethoxyXO4

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H20O3
CAS Number:
Molecular Weight:
344.40
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

, Faint yellow to dark orange

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

Biochem/physiol Actions

Methoxy-X04 (MeO-X04, MethoxyX04, MeX04, MX04) is a blood-brain barrier-permeant fluorescent probe derived from the Chrysamine G and Congo red scaffold that retains affinity toward amyloid beta (Aβ) fibrils (Ki = 25.3 nM/Chrysamine G and 26.8 nM/Methoxy-X04), while being much more lipophilic than Chrysamine G and Congo red. Methoxy-X04 stains Aβ plaques, tangles, and cerebrovascular amyloid in postmortem brain sections with good specificity and is commonly utilized for high-resolution fluorescent imaging of individual amyloid plaques in live animals by 2-photon microscopy (Ex. 750-800 nm; Em. 380-480, 500-540, and 560-650 nm) following i.v. or i.p. injection (5-10 mg/kg).

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kinetics of cerebral amyloid angiopathy progression in a transgenic mouse model of Alzheimer disease
The Journal of Neuroscience, 26(2), 365-371 (2006)
Mapping synaptic glutamate transporter dysfunction in vivo to regions surrounding A? plaques by iGluSnFR two-photon imaging
Nature Communications, 7, 13441-13441 (2016)
William E Klunk et al.
Journal of neuropathology and experimental neurology, 61(9), 797-805 (2002-09-17)
The identification of amyloid deposits in living Alzheimer disease (AD) patients is important for both early diagnosis and for monitoring the efficacy of newly developed anti-amyloid therapies. Methoxy-X04 is a derivative of Congo red and Chrysamine-G that contains no acid

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service