Skip to Content
Merck

SML3607

Etamsylate

≥95% (HPLC)

Synonym(s):

2,5-Dihydroxybenzenesulfonic acid diethylamine salt, CID 17506, Diethylammonium 2,5-dihydroxybenzenesulfonate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C6H6O5S·C4H11N
CAS Number:
Molecular Weight:
263.31
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

-10 to -25°C

Biochem/physiol Actions

Etamsylate is a haemostatic agent that is used for the management of blood loss. Apparently etamsylate increases capillary wall resistance and platelet adhesives by inhibition of the biosynthesis of prostaglandins causing platelet disaggregation in a presence of vascular wound. Recent studies suggest that etamsylate antagonizes anti-coagulant activity of heparin.
It is used for the management of blood loss; inhibits of the biosynthesis of prostaglandins causing platelet disaggregation

Disclaimer

Hygroscopic


Still not finding the right product?

Explore all of our products under Etamsylate


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library