Skip to Content
Merck

SML3319

WM-3835

≥98% (HPLC)

Synonym(s):

N′-(4-Fluoro-5-methylbiphenylcarbonyl)-3-hydroxybenzenesulfonohydrazide, WM 3835, WM3835, [1,1’-Biphenyl]-3-carboxylic acid, 4-fluoro-5-methyl-, 2-[(3-hydroxyphenyl)sulfonyl]hydrazide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H17FN2O4S
CAS Number:
Molecular Weight:
400.42
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

protect from light

color

white to beige

solubility

DMSO: soluble

storage temp.

−20°C

SMILES string

OC1=CC(S(NNC(C2=CC(C3=CC=CC=C3)=CC(C)=C2F)=O)(=O)=O)=CC=C1

InChI

1S/C20H17FN2O4S/c1-13-10-15(14-6-3-2-4-7-14)11-18(19(13)21)20(25)22-23-28(26,27)17-9-5-8-16(24)12-17/h2-12,23-24H,1H3,(H,22,25)

InChI key

KVJFJJXCBRSCDY-UHFFFAOYSA-N

Biochem/physiol Actions

Cell-permeable, potent, selective, Ac-CoA competitive and reversible inhibitor of HBO1, KAT6A, and TIP60 acetyltransferases
WM-3835is a cell-permeable hydroxybenzenesulfonohydrazide compound that acts as a potent, selective, Ac-CoA competitive and reversible inhibitor of HBO1, KAT6A, and TIP60 acetyltransferases (IC50 = 0.030, 0.017 and 0.312 µM, respectively). WM-3835 is non-cytotoxic, dose-dependently, and rapidly decreases H3K14Ac levels (IC50 = 0.341 µM) and inhibits AML cells growth (IC50 = 0.297 µM in Molm13). Induces apoptosis, downregulates HOXA and IL-6 expression and up-regulates ALB secretion and delays hepatocyte and hMPC senescence. WM-1385 potently inhibits POS-1 xenograft growth in SCID mice (10 mg/kg, i.p.).


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library