Skip to Content
Merck
All Photos(1)

Key Documents

SML3150

Sigma-Aldrich

GSK5182

≥95% (HPLC)

Synonym(s):

(δZ)-δ-[[4-[2-(Dimethylamino)ethoxy]phenyl](4-hydroxyphenyl)methylene]benzenebutanol, 4-[(Z)-1-[4-(2-Dimethylaminoethyloxy)phenyl]-hydroxy-2-phenylpent-1-enyl]phenol, GSK 5182, GSK-5182

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H31NO3
CAS Number:
Molecular Weight:
417.54
MDL number:
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥95% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Wramed)

storage temp.

2-8°C

Biochem/physiol Actions

GSK5182 is an orally available 4-hydroxy-tamoxifen (4-OHT) analog and a high affinity estrogen receptor-related receptor γ (ERRγ) inverse agonist (68% inhibition at 1 µM by reporter assay) with 25-fold reduced affinity and little antagonistic activity toward ERRα (No inhibition against 100 nM estradiol-induced reporter activity up to 1 µM). GSK5182 effectively inhibits ERRγ-dependent gene expression and biological functions both in cultures (1-10 µM) and in various animal studies in vivo (10-80 mg/kg i.p. or p.o. in rats and mice).

Pictograms

Health hazard

Signal Word

Danger

Hazard Statements

Hazard Classifications

Lact. - Repr. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Kamalakannan Radhakrishnan et al.
International journal of molecular sciences, 21(19) (2020-10-02)
Bone morphogenetic protein 6 (BMP6) is a multifunctional growth factor involved in organ development and homeostasis. BMP6 controls expression of the liver hormone, hepcidin, and thereby plays a crucial role in regulating iron homeostasis. BMP6 gene transcriptional regulation in liver
Yoon Seok Jung et al.
PloS one, 11(7), e0159425-e0159425 (2016-07-28)
Fibroblast growth factor 21 (FGF21), a stress inducible hepatokine, is synthesized in the liver and plays important roles in glucose and lipid metabolism. However, the mechanism of hepatic cannabinoid type 1 (CB1) receptor-mediated induction of FGF21 gene expression is largely
Hyo In Kim et al.
Pharmacological research, 165, 105423-105423 (2021-01-13)
Brain derived neurotrophic factor (BDNF) promotes maturation of dopaminergic (DAergic) neurons in the midbrain and positively regulates their maintenance and outgrowth. Therefore, understanding the mechanisms regulating the BDNF signaling pathway in DAergic neurons may help discover potential therapeutic strategies for
Yunhui Min et al.
Pharmaceuticals (Basel, Switzerland), 13(12) (2020-12-03)
Estrogen-related receptors (ERRs) are the first identified orphan nuclear receptors. The ERR family consists of ERRα, ERRβ, and ERRγ, regulating diverse isoform-specific functions. We have reported the importance of ERRγ in osteoarthritis (OA) pathogenesis. However, therapeutic approaches with ERRγ against
Juhee Lim et al.
Neurochemistry international, 136, 104726-104726 (2020-03-17)
Midbrain dopaminergic (DAergic) neurotransmission plays a crucial role in regulating motor, cognitive, and emotional functions. The orphan nuclear receptor estrogen-related receptor gamma (ERRγ) is highly expressed in the adult brain and in the developing fetal brain. Our previous study showed

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service