Skip to Content
Merck

SML2532

Jedi2

≥98% (HPLC)

Synonym(s):

2-Methyl-5-(2-thienyl)-3-furancarboxylic acid

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C10H8O3S
CAS Number:
Molecular Weight:
208.23
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[s]1c(ccc1)c2[o]c(c(c2)C(=O)O)C

InChI

1S/C10H8O3S/c1-6-7(10(11)12)5-8(13-6)9-3-2-4-14-9/h2-5H,1H3,(H,11,12)

InChI key

YXDCRSXNEOKXDE-UHFFFAOYSA-N

Biochem/physiol Actions

Jedi2 is an activator of the mechanosensitive Piezo1 channel that binds at a different place from Yoda1. It exhibits long-range allosteric gating, activating Piezo1 through the extracellular side of its peripheral blade-like structure rather than the C-terminal extracellular domain of the pore.
Piezo1 channel activator


pictograms

Exclamation mark

signalword

Warning

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library