Skip to Content
Merck

SML2449

THIQ

≥98% (HPLC)

Synonym(s):

(3R)-N-[(1R)-1-[(4-chlorophenyl)methyl]-2-[4-cyclohexyl-4-(1H-1,2,4-triazol-1-ylmethyl)-1-piperidinyl]-2-oxoethyl]-1,2,3,4-tetrahydro-3-isoquinolinecarboxamide, (3R)-N-[(2R)-3-(4-chlorophenyl)-1-[4-cyclo-hexyl-4-(1,2,4-triazol-1-ylmethyl)piperidin-1-yl]-1-oxopropan-2-yl]-1,2,3,4-tetrahydroisoquinoline-3-carboxamide

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C33H41ClN6O2
CAS Number:
Molecular Weight:
589.17
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C(N1CCC(CN2N=CN=C2)(C3CCCCC3)CC1)[C@@H](CC4=CC=C(Cl)C=C4)NC([C@@H]5NCC6=CC=CC=C6C5)=O

InChI

1S/C33H41ClN6O2/c34-28-12-10-24(11-13-28)18-30(38-31(41)29-19-25-6-4-5-7-26(25)20-36-29)32(42)39-16-14-33(15-17-39,21-40-23-35-22-37-40)27-8-2-1-3-9-27/h4-7,10-13,22-23,27,29-30,36H,1-3,8-9,14-21H2,(H,38,41)/t29-,30-/m1/s1

InChI key

HLCHESOMJVGDSJ-LOYHVIPDSA-N

Biochem/physiol Actions

THIQ is a selective tetrahydroisoquinoline melanocortin-4 receptor (MC4R) agonist. It exhibited an IC50 vaue of 1.2 nM in MC4R human receptors expressed in CHO cells compared values of 2067, 761 and 326 nM for MC1R, MC3R, and MC5R receptors, respectively.
Tetrahydroisoquinoline melanocortin-4 receptor (MC4R) agonist


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library