Skip to Content
Merck
All Photos(1)

Key Documents

SML1994

Sigma-Aldrich

BOP

>97% (HPLC)

Synonym(s):

N-(Benzenesulfonyl)-L-prolyl-L-O-(1-pyrrolidinylcarbonyl)tyrosine sodium salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H28N3NaO7S
CAS Number:
Molecular Weight:
537.56
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

>97% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 2 mg/mL, clear (warmed)

storage temp.

2-8°C

Biochem/physiol Actions

BOP (N-(Benzenesulfonyl)-L-prolyl-L-O-(1-pyrrolidinylcarbonyl)tyrosine sodium salt) is a potent and selective dual inhibitor of integrins alpha-4 beta-1 (α4β1) and alpha-9 beta-1 (α9β1) with Kd values in the picomolar range. BOP has been shown to preferentially mobilize hematopoietic stem cells (HSCs) and progenitors and to act synergistically with AMD3100.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Benjamin Cao et al.
Nature communications, 7, 11007-11007 (2016-03-16)
The inherent disadvantages of using granulocyte colony-stimulating factor (G-CSF) for hematopoietic stem cell (HSC) mobilization have driven efforts to identify alternate strategies based on single doses of small molecules. Here, we show targeting α9β1/α4β1 integrins with a single dose of
R Blake Pepinsky et al.
Biochemistry, 41(22), 7125-7141 (2002-05-30)
Integrins alpha9beta1 and alpha4beta1 form a distinct structural class, but while alpha4beta1 has been subjected to extensive study, alpha9beta1 remains poorly characterized. We have used the small molecule N-(benzenesulfonyl)-(L)-prolyl-(L)-O-(1-pyrrolidinylcarbonyl)tyrosine (3) to investigate the biochemical properties of alpha9beta1 and directly compare

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service