Skip to Content
Merck

SML1848

PCS1055 dihydrochloride

≥98% (HPLC)

Synonym(s):

6,7-Dihydro-N-[2-[1-(phenylmethyl)-4-piperidinyl]ethyl]-5H-benzo[6,7]cyclohepta[1,2-c]pyridazin-3-amine dihydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C27H32N4 · 2HCl
CAS Number:
Molecular Weight:
485.49
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

H2O: 5 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[H]Cl.[H]Cl.C12=C(N=NC(NCCC3CCN(CC4=CC=CC=C4)CC3)=C2)C5=C(C=CC=C5)CCC1

InChI

1S/C27H32N4.2ClH/c1-2-7-22(8-3-1)20-31-17-14-21(15-18-31)13-16-28-26-19-24-11-6-10-23-9-4-5-12-25(23)27(24)30-29-26;;/h1-5,7-9,12,19,21H,6,10-11,13-18,20H2,(H,28,29);2*1H

InChI key

VSCSFYDNGYAWKG-UHFFFAOYSA-N

Biochem/physiol Actions

PCS1055 is a potent and selective muscarinic M4 acetylcholine receptor competitive antagonist that exhibits >100-fold selectivity over M1-, M3-, and M5-receptors and 30-fold selectivity at the M2 receptor. PCS1055 was originally described as a potent electric eel AChE inhibitor (IC50 = 22 nM) and less potent at human AChE (IC50 = 120 nM).
Potent and selective muscarinic M4 receptor antagonist


hcodes

Hazard Classifications

Aquatic Chronic 4

Storage Class

11 - Combustible Solids

wgk

WGK 3



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library