Skip to Content
Merck

SML1829

ADX71743

≥98% (HPLC)

Synonym(s):

6-(2,4-Dimethylphenyl)-2-ethyl-6,7-dihydrobenzo[d]oxazol-4(5H)-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C17H19NO2
CAS Number:
Molecular Weight:
269.34
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
eCl@ss:
32160701
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 5 mg/mL, clear (warmed)

storage temp.

−20°C

SMILES string

CC1=CC(C)=CC=C1C2CC(OC(CC)=N3)=C3C(C2)=O

InChI key

CPKZCQHJDFSOJT-UHFFFAOYSA-N

Biochem/physiol Actions

ADX71743 is a brain-penetrant, selective and potent negative allosteric modulator of metabotropic glutamate receptor 7 (mGlu7).
ADX71743 is a brain-penetrant, selective and potent negative allosteric modulator of metabotropic glutamate receptor 7 (mGlu7). In one study ADX71743 exhibited an anxiolytic-like profile in rat and mouse models without causing impairment of locomotor activity. In another study it induced absence epileptic seizures and lethargy.
ADX71743 is known to negatively regulate the effect of mGlu7 (metabotropic glutamate receptor subtype 7), which is associated with a number of anxiety disorders.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library