Skip to Content
Merck

SML1685

Ribocil-C

≥98% (HPLC), Riboflavin riboswitch inhibitor, powder

Synonym(s):

2-[(3S)-1-[[1-(2-Pyrimidinyl)-1H-imidazol-4-yl]methyl]-3-piperidinyl]-6-(2-thienyl)-4(3H)-pyrimidinone

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C21H21N7OS
CAS Number:
Molecular Weight:
419.50
UNSPSC Code:
12352200
NACRES:
NA.77
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Product Name

Ribocil-C, ≥98% (HPLC)

Quality Segment

assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 20 mg/mL, clear

storage temp.

2-8°C

SMILES string

O=C1NC([C@@]2([H])CCCN(CC3=CN(C4=NC=CC=N4)C=N3)C2)=NC(C5=CC=CS5)=C1

Biochem/physiol Actions

Ribocil-C is a potent inhibitor of bacterial riboflavin riboswitches.
Ribocil-C is a potent inhibitor of bacterial riboflavin riboswitches. Riboswitches are cis regulatory elements present in non-coding RNA that specifically bind to natural ligands to regulate gene expression. Ribocil-C blocks the flavin mononucleotide riboswitch-mediated expression of the ribB gene, which is required for riboflavin biosynthesis, with an IC50 value of 23 nM. Ribocil-C inhibited bacterial cell growth of E. coli MB5746 and bacterial burden in a mouse E. coli septicaemia model of infection.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library