Skip to Content
Merck

SML1576

Undecylprodigiosin hydrochloride

≥95% (HPLC)

Synonym(s):

2-[(Z)-[3-Methoxy-5-(1H-pyrrol-2-yl)-2H-pyrrol-2-ylidene]methyl]-5-undecyl-1H-pyrrole hydrochloride, CID 6438379 hydrochloride, Prodigiosin C-25 hydrochloride, Undecylprodiginin hydrochloride, Undecylprodiginine hydrochloride

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C25H35N3O · HCl
CAS Number:
Molecular Weight:
430.03
UNSPSC Code:
12352200
NACRES:
NA.77
Assay:
≥95% (HPLC)
Form:
powder
Storage condition:
desiccated, protect from light
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


biological source

Streptomyces parvulus

Quality Segment

assay

≥95% (HPLC)

form

powder

storage condition

desiccated, protect from light

color

, faint red to dark red

storage temp.

−20°C

SMILES string

COC1=CC(C2=CC=CN2)=N/C1=C\C3=CC=C(CCCCCCCCCCC)N3.[H]Cl

InChI

1S/C25H35N3O.ClH/c1-3-4-5-6-7-8-9-10-11-13-20-15-16-21(27-20)18-24-25(29-2)19-23(28-24)22-14-12-17-26-22;/h12,14-19,26-27H,3-11,13H2,1-2H3;1H/b24-18-;

InChI key

ZZRPZWXEPLULCE-XWASNXTISA-N

Biochem/physiol Actions

Undecylprodigiosin is a tri-pyrolle antibiotic isolated from Streptomyces coelicolor A3 that exhibits immunosuppressive and apoptotic activities as well as antibacterial, antioxidative and UV protective properties.
Undecylprodigiosin is a tri-pyrolle antibiotic isolated from Streptomyces coelicolor A3 that exhibits immunosuppressive and apoptotic activities as well as antibacterial, antioxidative and UV protective properties. Undecylprodigiosin selectively induces p53 independent apoptosis in cancer cells.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library