Skip to Content
Merck

SML1506

Sigma-Aldrich

KY-05009

≥98% (HPLC)

Synonym(s):

5-(4-Methylbenzamido)-2-(phenylamino)thiazole-4-carboxamide, 5-[(4-Methylbenzoyl)amino]-2-(phenylamino)-4-thiazolecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H16N4O2S
CAS Number:
Molecular Weight:
352.41
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Pricing and availability is not currently available.

Quality Level

Assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

NC(C1=C(NC(C2=CC=C(C)C=C2)=O)SC(NC3=CC=CC=C3)=N1)=O

InChI

1S/C18H16N4O2S/c1-11-7-9-12(10-8-11)16(24)22-17-14(15(19)23)21-18(25-17)20-13-5-3-2-4-6-13/h2-10H,1H3,(H2,19,23)(H,20,21)(H,22,24)

InChI key

WCEDGRTWDSHZHF-UHFFFAOYSA-N

Biochem/physiol Actions

KY-05009 is a Traf2- and Nck-interacting kinase (TNIK) inhibitor that inhibits the epithelial-to-mesenchymal transition (EMT) in cancer cells.
KY-05009 is also known as (5-(4-methylbenzamido)-2-(phenylamino) thiazole-4-carboxamide). It prevents the proliferation induced by interleukin-6 and promotes cell death by blocking Wnt signaling in multiple myeloma (MM) cells.[1] KY-05009 is a novel aminothiazole, which possesses anti-cancer activity in human colorectal cancer cells. KY-05009 prevents migration and invasion stimulated by TGF-β1 (transforming growth factor beta 1).[1]

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Synergistic inhibition effect of TNIK inhibitor KY-05009 and receptor tyrosine kinase inhibitor dovitinib on IL-6-induced proliferation and Wnt signaling pathway in human multiple myeloma cells
Lee Y, et al.
Oncotarget, 8(25), 41091-41091 (2017)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service