Skip to Content
Merck

Skip To

SML1292

TAPI-0

≥95% (HPLC)

Synonym(s):

N-(R)-(2-(Hydroxyaminocarbonyl)Methyl)-4-Methylpentanoyl-L-Naphthylalanyl-L-Alanine amide, TNF-α Processing Inhibitor-0, TNF-α Protease Inhibitor-0

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C24H32N4O5
CAS Number:
Molecular Weight:
456.53
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥95% (HPLC)
Form:
powder
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥95% (HPLC)

form

powder

color

white to off-white

shipped in

wet ice

storage temp.

−20°C

SMILES string

NC([C@@](C)([H])NC([C@]([H])(CC1=CC=CC2=C1C=CC=C2)NC([C@@](CC(C)C)([H])CC(NO)=O)=O)=O)=O

InChI

1S/C24H32N4O5/c1-14(2)11-18(13-21(29)28-33)23(31)27-20(24(32)26-15(3)22(25)30)12-17-9-6-8-16-7-4-5-10-19(16)17/h4-10,14-15,18,20,33H,11-13H2,1-3H3,(H2,25,30)(H,26,32)(H,27,31)(H,28,29)/t15-,18+,20-/m0/s1

InChI key

WSJJYJNVFYARGB-CVAIRZPRSA-N

Biochem/physiol Actions

TAPI-0 is a metalloproteinase inhibitor with selectivity for TACE (TNF-α converting enzyme/ADAM17; IC50 = 100 nM). TAPI-0 also inhibits of collagenase and gelatinase. Although not specific for TACE, TAPI-0 is extensively used to block TACE-mediated TNF-α shedding.
TAPI-0 is a metalloproteinase inhibitor with selectivity for TACE; also inhibits collagenase and gelatinase.
TAPI-0 inhibit peptide deformylase (PDF) activity and retard the growth Chlamydia trachomatis, which is an obligate intracellular bacterium responsible for a number of human diseases.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
579051579050579052
form

powder

form

solid

form

solid

form

solid

assay

≥95% (HPLC)

assay

≥97% (HPLC)

assay

≥96% (HPLC)

assay

≥97% (HPLC)

Quality Level

100

Quality Level

100

Quality Level

100

Quality Level

100

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

storage temp.

−20°C

color

white to off-white

color

white

color

white

color

white

shipped in

wet ice

shipped in

ambient

shipped in

ambient

shipped in

ambient


pictograms

Skull and crossbones

signalword

Danger

hcodes

Hazard Classifications

Acute Tox. 3 Oral

Storage Class

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters