Skip to Content
Merck

Skip To

SML0549

JTV-519 hemifumarate

≥98% (HPLC)

Synonym(s):

1-(2,3-Dihydro-7-methoxy-1,4-benzothiazepin-4(5H)-yl)-3-[4-(phenylmethyl)-1-piperidinyl]-1-propanone hemifumarate, K201 hemifumarate

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View
Size/SKUAvailabilityPrice

About This Item

Empirical Formula (Hill Notation):
C25H32N2O2S · .5C4H4O4
CAS Number:
Molecular Weight:
482.63
NACRES:
NA.77
PubChem Substance ID:
UNSPSC Code:
12352200
MDL number:
Assay:
≥98% (HPLC)
Form:
powder
Storage condition:
desiccated
Pricing and availability is not currently available.
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 10 mg/mL, clear

storage temp.

2-8°C

SMILES string

OC(=O)\C=C\C(O)=O.COc1ccc2SCCN(Cc2c1)C(=O)CCN3CCC(CC3)Cc4ccccc4.COc5ccc6SCCN(Cc6c5)C(=O)CCN7CCC(CC7)Cc8ccccc8

InChI

1S/2C25H32N2O2S.C4H4O4/c2*1-29-23-7-8-24-22(18-23)19-27(15-16-30-24)25(28)11-14-26-12-9-21(10-13-26)17-20-5-3-2-4-6-20;5-3(6)1-2-4(7)8/h2*2-8,18,21H,9-17,19H2,1H3;1-2H,(H,5,6)(H,7,8)/b;;2-1+

InChI key

MNAIJOCDPCXVEY-WXXKFALUSA-N

Biochem/physiol Actions

JTV-519 is a RYR2 antagonist.
JTV-519 is a potent inhibitor or the ryanodine receptor 2 (RYR2) blocker. RYR2 is a cardiac calcium channel that regulates calcium levels in the sarcoplasmic reticulum. JTV-519 stabilizes RYR2 in the closed state.

Features and Benefits

This compound is featured on the InsP3/Ryanodine Receptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.

Compare Similar Items

View Full Comparison

Show Differences

1 of 1

This Item
SML2541S7201B2185
assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

assay

≥98% (HPLC)

form

powder

form

powder

form

powder

form

solid

Quality Level

100

Quality Level

-

Quality Level

100

Quality Level

100

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

2-8°C

storage temp.

-

storage condition

desiccated

storage condition

-

storage condition

protect from light

storage condition

-

solubility

DMSO: 10 mg/mL, clear

solubility

DMSO: 2 mg/mL, clear

solubility

DMSO: ≥10 mg/mL

solubility

DMSO: >20 mg/mL


pictograms

Environment

signalword

Warning

hcodes

Hazard Classifications

Aquatic Acute 1 - Aquatic Chronic 1

Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library



Questions

Reviews

No rating value

Active Filters