Skip to Content
Merck

SML0176

Z36

≥98% (HPLC)

Synonym(s):

(Z)-5-Fluoro-1-(3′-dimethylamino)propyl-3-[(5′-methoxyindol-3-ylidene)methyl]-indolin-2-one, 1-[3-(dimethylamino)propyl]-5-fluoro-1,3-dihydro-3-[(5-methoxy-1H-indol-3-yl)methylene]-2H-Indol-2-one

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C23H24FN3O2
CAS Number:
Molecular Weight:
393.45
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Assay:
≥98% (HPLC)
Form:
powder
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


Quality Segment

assay

≥98% (HPLC)

form

powder

color

light yellow to dark yellow

solubility

DMSO: ≥15 mg/mL

storage temp.

−20°C

SMILES string

COc1ccc2[nH]cc(\C=C3/C(=O)N(CCCN(C)C)c4ccc(F)cc34)c2c1

InChI

1S/C23H24FN3O2/c1-26(2)9-4-10-27-22-8-5-16(24)12-19(22)20(23(27)28)11-15-14-25-21-7-6-17(29-3)13-18(15)21/h5-8,11-14,25H,4,9-10H2,1-3H3/b20-11-

InChI key

CQNOITUXWKRVMB-JAIQZWGSSA-N

Application

Z36 may be used to study cell signaling that results in autophagy.

Biochem/physiol Actions

Bcl-XL inhibitor that induces autophagic cell death independent of apoptosis
Z36 inhibits cell proliferation and induces death in HeLa cells. The cell death induced is via autophagy and autophagic flux.
Z36 is a Bcl-XL inhibitor that induces autophagic cell death independent of apoptosis which distinguishes it from other Bcl-XL and Bcl-2 inhibitors that induce cancer cell death mainly through apoptosis. Z36 binds to the BH3 binding groove of BCL-XL where it is thought to induce autophagy through blocking the interaction between Bcl-XL/Bcl2 and Beclin 1.


Storage Class

11 - Combustible Solids

wgk

WGK 3

flash_point_f

Not applicable

flash_point_c

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library