SMB01083
4,5-Di-O-caffeoylquinic acid methyl ester
≥90% (LC/MS-ELSD)
Synonym(s):
Methyl (1R,3R,4S,5R)-3,4-bis[[(E)-3-(3,4-dihydroxyphenyl)prop-2-enoyl]oxy]-1,5-dihydroxycyclohexane-1-carboxylate
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
biological source
plant
Assay
≥90% (LC/MS-ELSD)
form
solid
mol wt
530.48
application(s)
metabolomics
vitamins, nutraceuticals, and natural products
storage temp.
−20°C
General description
4,5-Di-O-caffeoylquinic acid methyl ester, a phenylpropanoid, belongs to group of compounds known as cyclic alcohols and derivatives. It is a natural product commonly available from Erigeron breviscapus, Spuriopimpinella brachycarpa, Ipomoea batatas L., and Spuriopimpinella calycina plants. Studies have demonstrated that this metabolite, derived from plant sources, possesses various biological activities, including antioxidant and antiviral properties.
Application
It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and pharmaceutical research.
Biochem/physiol Actions
4,5-Di-O-caffeoylquinic acid methyl ester demonstrates strong antioxidant activity with significant free radical-scavenging abilities against DPPH. Additionally, it displays in vitro antiviral activity against the respiratory syncytial virus.
Features and Benefits
- High quality compound suitable for multiple research applications
- Compatible with HPLC and mass spectrometry techniques
Other Notes
For additional information on our range of Biochemicals, please complete this form.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Phytomedicine : international journal of phytotherapy and phytopharmacology, 70, 153219-153219 (2020-05-04)
4,5-di-O-caffeoylquinic acid methyl ester (4,5-CQME) is a caffeoylquinic acid (CQA) isolated from Lonicera japonica Thunb., a traditional Chinese medicine. To date, the biological activity of 4,5-CQME has not been fully investigated. The aim of the current study was to explore
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service