Skip to Content
Merck

SMB00569

2′,4′-Dihydroxy-4-prenyloxychalcone

≥85% (LC/MS-UV)

Synonym(s):

LMPK12120217

Sign In to View Organizational & Contract Pricing.

Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C20H20O4
CAS Number:
Molecular Weight:
324.37
NACRES:
NA.25
PubChem Substance ID:
UNSPSC Code:
12352205
MDL number:
Technical Service
Need help? Our team of experienced scientists is here for you.
Let Us Assist


assay

≥85% (LC/MS-UV)

form

solid

solubility

DMSO: 1 mg/mL

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O=C(C1=C(C=C(C=C1)O)O)/C=C/C2=CC=C(OCC=C(C)C)C=C2

InChI

1S/C20H20O4/c1-14(2)11-12-24-17-7-3-15(4-8-17)5-10-19(22)18-9-6-16(21)13-20(18)23/h3-11,13,21,23H,12H2,1-2H3/b10-5+

InChI key

VVSZRCIUFBADJQ-BJMVGYQFSA-N

General description

Natural product derived from plant source.


Storage Class

11 - Combustible Solids

WGK

WGK 3

Flash Point (°F)

Not applicable

Flash Point (°C)

Not applicable



Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library